Assay Details
| Gene Symbol: Tead1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166584.1 | NP_001160056.1 | 6 - 7 | 870 | 83 | |
| NM_009346.3 | NP_033372.1 | 5 - 6 | 807 | 83 | ||
| GenBank mRNA | AK140307.1 | - | 6 - 7 | 871 | 83 | |
| AK148375.1 | - | 5 - 6 | 773 | 83 | ||
| L13853.1 | - | 5 - 6 | 534 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tead1
Genomic Map