Assay Details
| Gene Symbol: Stard9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK029170.1 | - | 1819 | 88 | ||
| AK029732.1 | - | 7 - 8 | 3168 | 88 | ||
| AK122489.1 | - | 7 - 8 | 3868 | 88 | ||
| BC032885.1 | - | 5 - 6 | 578 | 88 | ||
More Information
Set Membership: |
|

Stard9
Genomic Map