Assay Details
| Gene Symbol: Ccndbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010761.2 | NP_034891.2 | 8 - 9 | 952 | 77 | |
| GenBank mRNA | AK144883.1 | - | 8 - 9 | 929 | 77 | |
| AK154285.1 | - | 8 - 9 | 906 | 77 | ||
| AK160205.1 | - | 8 - 9 | 918 | 77 | ||
| AK163150.1 | - | 8 - 9 | 865 | 77 | ||
| AK167345.1 | - | 8 - 9 | 917 | 77 | ||
| BC001984.1 | - | 8 - 9 | 888 | 3489973 | 77 | |
| BC004673.1 | - | 2 - 3 | 372 | 77 | ||
| BC005802.1 | - | 7 - 8 | 961 | 3707919 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccndbp1
Genomic Map