Assay Details
| Gene Symbol: Slc22a18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042760.1 | NP_001036225.1 | 3 - 4 | 459 | 110 | |
| NM_008767.2 | NP_032793.2 | 3 - 4 | 419 | 110 | ||
| GenBank mRNA | AB012084.1 | - | 3 - 4 | 459 | 110 | |
| AF028739.1 | - | 3 - 4 | 375 | 110 | ||
| AK002460.1 | - | 3 - 4 | 400 | 110 | ||
| BC003734.1 | - | 3 - 4 | 365 | 3599736 | 110 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc22a18
Genomic Map