Assay Details
| Gene Symbol: Slc22a18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042760.1 | NP_001036225.1 | 4 - 5 | 620 | 61 | |
| NM_008767.2 | NP_032793.2 | 4 - 5 | 580 | 61 | ||
| GenBank mRNA | AB012084.1 | - | 4 - 5 | 620 | 61 | |
| AF028739.1 | - | 4 - 5 | 536 | 61 | ||
| AK002460.1 | - | 4 - 5 | 561 | 61 | ||
| BC003734.1 | - | 4 - 5 | 526 | 3599736 | 61 | |
| BC015078.1 | - | 3 - 4 | 467 | 4224844 | 61 | |
| BE865039.1 | - | 1 - 2 | 87 | 61 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC015078.1 transcripts.
Set Membership: |
|

Slc22a18
Genomic Map