Assay Details
| Gene Symbol: Slc22a18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042760.1 | NP_001036225.1 | 6 - 7 | 861 | 66 | |
| NM_008767.2 | NP_032793.2 | 6 - 7 | 821 | 66 | ||
| GenBank mRNA | AB012084.1 | - | 6 - 7 | 861 | 66 | |
| AF037065.1 | - | 6 - 7 | 792 | 66 | ||
| BC003734.1 | - | 6 - 7 | 767 | 3599736 | 66 | |
| BC015078.1 | - | 5 - 6 | 708 | 4224844 | 66 | |
| BE865039.1 | - | 3 - 4 | 328 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc22a18
Genomic Map