Assay Details
| Gene Symbol: Slc22a18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042760.1 | NP_001036225.1 | 7 - 8 | 972 | 96 | |
| NM_008767.2 | NP_032793.2 | 7 - 8 | 932 | 96 | ||
| GenBank mRNA | AB012084.1 | - | 7 - 8 | 972 | 96 | |
| AF028739.1 | - | 7 - 8 | 888 | 96 | ||
| AK002460.1 | - | 7 - 8 | 914 | 96 | ||
| BC003734.1 | - | 7 - 8 | 878 | 3599736 | 96 | |
| BC015078.1 | - | 6 - 7 | 819 | 4224844 | 96 | |
| BE865039.1 | - | 4 - 5 | 439 | 96 | ||
More Information
Set Membership: |
|

Slc22a18
Genomic Map