Assay Details
| Gene Symbol: Slc22a18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042760.1 | NP_001036225.1 | 8 - 9 | 1072 | 72 | |
| NM_008767.2 | NP_032793.2 | 8 - 9 | 1032 | 72 | ||
| GenBank mRNA | AB012084.1 | - | 8 - 9 | 1072 | 72 | |
| AF028739.1 | - | 8 - 9 | 988 | 72 | ||
| AF037065.1 | - | 8 - 9 | 1000 | 72 | ||
| BC003734.1 | - | 8 - 9 | 978 | 3599736 | 72 | |
| BC015078.1 | - | 7 - 8 | 919 | 4224844 | 72 | |
| BE865039.1 | - | 5 - 6 | 539 | 72 | ||
| BU758157.1 | - | 3 - 4 | 72 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Slc22a18
Genomic Map