Assay Details
| Gene Symbol: Rad51ap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111118.1 | NP_001104588.1 | 2 - 3 | 2813 | 151 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1999862 | NM_001111118.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 823654 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 628353 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 545646 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 124165 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 948771 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 166753 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1999862 | NM_001111118.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P357948 | NM_001111118.1 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238697 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28238713 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28238681 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P10432643 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28238713 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28238625 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28238699 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28238694 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28238632 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28238695 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28238711 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28238709 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28238628 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28238623 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P10262857 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28238631 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28238690 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28238670 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28238702 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28238666 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28238661 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28238674 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28238672 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28238714 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28238649 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28238706 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28238629 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28238651 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28238636 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P10058801 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28238703 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28238677 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28238646 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28238657 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28238679 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28238668 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28238683 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28238627 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28238707 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28238639 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28238701 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28238652 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28238693 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28238684 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28238659 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28238633 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28238644 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28238686 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28238689 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28238665 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28238655 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28238642 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28238662 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Clariom S Array | PSR1200000953.mm.1 | NM_001111118.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200000953.mm.1 | NM_001111118.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1200000953.mm.1 | TC1200000093.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 453321 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1248183 | - | - |

Rad51ap2
Genomic Map