Assay Details
| Gene Symbol: Gm38407 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110487.1 | 10 - 11 | 1892 | 107 | ||
| GenBank mRNA | AK029853.1 | - | 10 - 11 | 1894 | 107 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | retinol dehydrogenase 14 (all-trans and 9-cis) | Rdh14 |
3110030G19Rik, AW215439, PAN2 |
-117.3kb | Sense | Chr. 12 - 10390690 - 10395562 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 7420701I03 gene | 7420701I03Rik | 533.0kb | Sense | Chr. 12 - 11047813 - 11078741 on NCBI Build GRCm38 |
More Information
Set Membership: |
|
MicroArray Probes:
Agilent
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 281148 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 929099 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 684237 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 306006 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 711100 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 819953 | AK035480.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 487790 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 618845 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 120693 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 712221 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 151497 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 312219 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 880045 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 32340 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 364648 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 641227 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 274447 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 168776 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 517674 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 578991 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 263922 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 150251 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 320777 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 737477 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 79451 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 500484 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 776364 | BC016539.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 776364 | NM_001024720.3 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 322781 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 160869 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 37694 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 96501 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 176307 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 120138 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 129202 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07537227 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07537268 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02459939 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07537164 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07537245 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07537138 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07537110 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07537196 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P102538 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P07537196 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P07537245 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P07537110 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28233500 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28233755 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28234063 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28234091 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28234082 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28233536 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28233753 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28233502 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28234066 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28233593 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28234057 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28234084 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28233757 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28234060 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28234056 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28233496 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28233756 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28233739 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28233600 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28233513 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28233498 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28233530 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28233493 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P10189776 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28234077 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28233510 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09893812 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28233520 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28233601 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28233748 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28233603 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28234098 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28234073 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28234069 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28233528 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28233742 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28233597 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28233533 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28233529 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28233495 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28234097 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28234088 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28234054 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28233750 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28234078 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28233532 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28234059 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28234067 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28233740 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P10184929 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28234094 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28233516 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28233508 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28233509 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28233605 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28234081 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28234095 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28234049 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28233745 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28233598 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09761924 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28233734 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28233584 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28233512 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28233504 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28234058 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28234065 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28233585 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28234051 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28233747 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28233592 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P10039375 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28233594 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28233752 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28234093 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28234076 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28234068 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28234100 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28233523 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28233588 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28233736 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28234072 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28233507 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28233525 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28234085 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28234071 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28234052 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 660024 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 100430528 | - | - |

Gm38407
Genomic Map