Assay Details
| Gene Symbol: Pax6os1 | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002867.2 | 2 - 3 | 197 | 58 | |||
| GenBank mRNA | AK029183.1 | - | LIT3400 Pax6OS |
2 - 3 | 198 | 58 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | reticulocalbin 1 | Rcn1 | Rcn | -185.5kb | Sense | Chr. 2 - 105385948 - 105399319 on NCBI Build GRCm38 |
| Downstream | Pax6 upstream antisense RNA | Paupar | 59.5kb | Sense | Chr. 2 - 105657862 - 105661343 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Pax6os1
Genomic Map