Assay Details
| Gene Symbol: Gyltl1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166633.2 | NP_001160105.1 | 10 - 11 | 1265 | 69 | |
| NM_001290773.1 | NP_001277702.1 | 11 - 12 | 1373 | 69 | ||
| NM_001290774.1 | NP_001277703.1 | 10 - 11 | 1287 | 69 | ||
| NM_001290775.1 | NP_001277704.1 | 11 - 12 | 1590 | 69 | ||
| NM_172670.3 | NP_766258.2 | 11 - 12 | 1370 | 69 | ||
| GenBank mRNA | AK077630.1 | - | 10 - 11 | 1241 | 69 | |
| AY742914.1 | - | 11 - 12 | 1346 | 69 | ||
| AY742915.1 | - | 10 - 11 | 1241 | 69 | ||
| BC058199.1 | - | 11 - 12 | 1551 | 69 | ||
| BC096655.1 | - | 10 - 11 | 1297 | 5057489 | 69 | |
| BC116715.1 | - | 10 - 11 | 1302 | 40093557 | 69 | |
| HM141800.1 | - | 10 - 11 | 1199 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gyltl1b
Genomic Map