Assay Details
| Gene Symbol: Gyltl1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166633.2 | NP_001160105.1 | 11 - 12 | 1545 | 85 | |
| NM_001290773.1 | NP_001277702.1 | 12 - 13 | 1653 | 85 | ||
| NM_001290774.1 | NP_001277703.1 | 11 - 12 | 1567 | 85 | ||
| NM_001290775.1 | NP_001277704.1 | 12 - 13 | 1870 | 85 | ||
| NM_172670.3 | NP_766258.2 | 12 - 13 | 1650 | 85 | ||
| GenBank mRNA | AK077630.1 | - | 11 - 12 | 1521 | 85 | |
| AK220181.1 | - | 3 - 4 | 3989 | 85 | ||
| AY742914.1 | - | 12 - 13 | 1626 | 85 | ||
| AY742915.1 | - | 11 - 12 | 1521 | 85 | ||
| BC004023.1 | - | 1 - 2 | 255 | 85 | ||
| BC033922.1 | - | 2 - 3 | 303 | 85 | ||
| BC058199.1 | - | 12 - 13 | 1831 | 85 | ||
| BC096655.1 | - | 11 - 12 | 1577 | 5057489 | 85 | |
| BC116715.1 | - | 11 - 12 | 1582 | 40093557 | 85 | |
| HM141800.1 | - | 11 - 12 | 1479 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gyltl1b
Genomic Map