Assay Details
| Gene Symbol: Gyltl1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166633.2 | NP_001160105.1 | 5 - 6 | 541 | 82 | |
| NM_001290773.1 | NP_001277702.1 | 5 - 6 | 544 | 82 | ||
| NM_001290774.1 | NP_001277703.1 | 4 - 5 | 458 | 82 | ||
| NM_001290775.1 | NP_001277704.1 | 5 - 6 | 761 | 82 | ||
| NM_172670.3 | NP_766258.2 | 5 - 6 | 541 | 82 | ||
| GenBank mRNA | AY742914.1 | - | 5 - 6 | 517 | 82 | |
| AY742915.1 | - | 5 - 6 | 517 | 82 | ||
| BC096655.1 | - | 4 - 5 | 468 | 5057489 | 82 | |
| BC116715.1 | - | 4 - 5 | 473 | 40093557 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gyltl1b
Genomic Map