Assay Details
| Gene Symbol: Gyltl1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166633.2 | NP_001160105.1 | 8 - 9 | 942 | 66 | |
| NM_001290773.1 | NP_001277702.1 | 9 - 10 | 1050 | 66 | ||
| NM_001290774.1 | NP_001277703.1 | 8 - 9 | 964 | 66 | ||
| NM_001290775.1 | NP_001277704.1 | 9 - 10 | 1267 | 66 | ||
| NM_172670.3 | NP_766258.2 | 9 - 10 | 1047 | 66 | ||
| GenBank mRNA | AK077630.1 | - | 8 - 9 | 918 | 66 | |
| AY742914.1 | - | 9 - 10 | 1023 | 66 | ||
| AY742915.1 | - | 8 - 9 | 918 | 66 | ||
| BC058199.1 | - | 9 - 10 | 1228 | 66 | ||
| BC096655.1 | - | 8 - 9 | 974 | 5057489 | 66 | |
| BC116715.1 | - | 8 - 9 | 979 | 40093557 | 66 | |
| HM141800.1 | - | 8 - 9 | 876 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gyltl1b
Genomic Map