Assay Details
| Gene Symbol: Mllt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010806.1 | NP_034936.1 | 10 - 11 | 1455 | 94 | |
| GenBank mRNA | AK016557.1 | - | 9 - 10 | 1574 | 94 | |
| CK637291.1 | - | 1 - 2 | 107 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mllt4
Genomic Map