Assay Details
| Gene Symbol: Mllt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010806.1 | NP_034936.1 | 11 - 12 | 1714 | 65 | |
| GenBank mRNA | AK016557.1 | - | 10 - 11 | 1833 | 65 | |
| CK637291.1 | - | 2 - 3 | 366 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mllt4
Genomic Map