Assay Details
| Gene Symbol: Mllt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010806.1 | NP_034936.1 | 16 - 17 | 2318 | 107 | |
| GenBank mRNA | AK016557.1 | - | 15 - 16 | 2437 | 107 | |
| CN537179.1 | - | 2 - 3 | 188 | 107 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mllt4
Genomic Map