Assay Details
| Gene Symbol: Mllt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010806.1 | NP_034936.1 | 18 - 19 | 2677 | 129 | |
| GenBank mRNA | AK016557.1 | - | 17 - 18 | 2796 | 129 | |
| CN537179.1 | - | 4 - 5 | 547 | 129 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mllt4
Genomic Map