Assay Details
| Gene Symbol: Mllt4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010806.1 | NP_034936.1 | 27 - 28 | 3760 | 80 | |
| GenBank mRNA | BU611478.1 | - | 3 - 4 | 362 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mllt4
Genomic Map