Assay Details
| Gene Symbol: Gucy2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130693.3 | NP_001124165.1 | 14 - 15 | 2884 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|
UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gucy2d| Gene Symbol: Gucy2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130693.3 | NP_001124165.1 | 14 - 15 | 2884 | 68 | |
Set Membership: |
|
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Clariom S Array | PSR0700011635.mm.1 | TC0700001366.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700011635.mm.1 | NM_001130693.3 | Refseq |
Genomic Map