Assay Details
| Gene Symbol: Gucy2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130693.3 | NP_001124165.1 | 7 - 8 | 1824 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gucy2d| Gene Symbol: Gucy2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130693.3 | NP_001124165.1 | 7 - 8 | 1824 | 66 | |
Set Membership: |
|