| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1700007575.mm.1 |
NM_001168502.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700007575.mm.1 |
NM_001168502.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700007575.mm.1 |
TC1700000752.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700007585.mm.1 |
NM_001013745.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700007585.mm.1 |
NM_001013745.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700007585.mm.1 |
NM_001168502.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700007585.mm.1 |
NM_001168502.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700007585.mm.1 |
TC1700000752.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700007576.mm.1 |
NM_001168502.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700007576.mm.1 |
NM_001168502.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700007576.mm.1 |
TC1700000752.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700007584.mm.1 |
NM_001013745.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700007584.mm.1 |
NM_001013745.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700007584.mm.1 |
NM_001168502.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700007584.mm.1 |
NM_001168502.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700007584.mm.1 |
TC1700000752.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700007589.mm.1 |
NM_001013745.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700007589.mm.1 |
NM_001013745.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700007589.mm.1 |
NM_001168501.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700007589.mm.1 |
NM_001168501.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700007589.mm.1 |
NM_001168502.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700007589.mm.1 |
NM_001168502.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700007589.mm.1 |
TC1700000752.mm.1 |
MTA TC |