| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1700007599.mm.1 |
NR_033137.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700007599.mm.1 |
NR_033137.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700007599.mm.1 |
TC1700000752.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700007577.mm.1 |
NM_001168501.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700007577.mm.1 |
NM_001168501.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700007577.mm.1 |
NM_001168502.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700007577.mm.1 |
NM_001168502.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700007577.mm.1 |
TC1700000752.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700007578.mm.1 |
NM_001013745.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700007578.mm.1 |
NM_001013745.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700007578.mm.1 |
NM_001168501.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700007578.mm.1 |
NM_001168501.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700007578.mm.1 |
NM_001168502.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700007578.mm.1 |
NM_001168502.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700007578.mm.1 |
TC1700000752.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700007596.mm.1 |
NR_033137.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700007596.mm.1 |
NR_033137.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700007596.mm.1 |
TC1700000752.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700007597.mm.1 |
NR_033137.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700007597.mm.1 |
NR_033137.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700007597.mm.1 |
TC1700000752.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700007598.mm.1 |
NR_033137.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700007598.mm.1 |
NR_033137.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700007598.mm.1 |
TC1700000752.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700007594.mm.1 |
NR_033137.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700007594.mm.1 |
NR_033137.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700007594.mm.1 |
TC1700000752.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700007604.mm.1 |
NM_001013745.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700007604.mm.1 |
NM_001013745.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700007604.mm.1 |
NM_001168501.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700007604.mm.1 |
NM_001168501.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700007604.mm.1 |
NM_001168502.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700007604.mm.1 |
NM_001168502.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700007604.mm.1 |
NR_033137.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700007604.mm.1 |
NR_033137.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700007604.mm.1 |
TC1700000752.mm.1 |
MTA TC |