Assay Details
| Gene Symbol: Ppif | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134084.1 | NP_598845.1 | 1 - 2 | 255 | 84 | |
| GenBank mRNA | AK028349.1 | - | 1 - 2 | 272 | 84 | |
| BC004041.1 | - | 1 - 2 | 255 | 3496285 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ppif
Genomic Map