Assay Details
| Gene Symbol: Tjap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252473.1 | NP_001239402.1 | 8 - 9 | 753 | 93 | |
| NM_001252474.1 | NP_001239403.1 | 8 - 9 | 750 | 93 | ||
| NM_001252475.1 | NP_001239404.1 | 7 - 8 | 656 | 93 | ||
| NM_028751.3 | NP_083027.1 | 9 - 10 | 883 | 93 | ||
| GenBank mRNA | AB712252.1 | - | 6 - 7 | 500 | 93 | |
| AK002882.2 | - | 9 - 10 | 870 | 93 | ||
| AK164925.1 | - | 8 - 9 | 736 | 93 | ||
| AK169889.1 | - | 8 - 9 | 729 | 93 | ||
| AK177408.1 | - | 3 - 4 | 272 | 94 | ||
| BC023316.1 | - | 9 - 10 | 869 | 3993257 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tjap1
Genomic Map