Assay Details
| Gene Symbol: Mrps18b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK002554.1 | - | 1 - 2 | 100 | 75 | |
| AK076033.1 | - | 1 - 2 | 100 | 75 | ||
| AK076044.1 | - | 1 - 2 | 100 | 75 | ||
More Information
Set Membership: |
|

Mrps18b
Genomic Map