Assay Details
| Gene Symbol: Unc5cl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK144925.1 | - | 1 - 2 | 65 | 64 | |
| BC095959.1 | - | 1 - 2 | 42 | 5349225 | 64 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK144925.1, BC095959.1 transcripts.
Set Membership: |
|

Unc5cl
Genomic Map