Assay Details
| Gene Symbol: Cchcr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146248.2 | NP_666360.2 | 11 - 12 | 1592 | 57 | |
| GenBank mRNA | AK159780.1 | - | 11 - 12 | 1519 | 57 | |
| BC031416.1 | - | 12 - 13 | 1681 | 4012768 | 57 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Cchcr1
Genomic Map