Assay Details
| Gene Symbol: Cchcr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146248.2 | NP_666360.2 | 1 - 2 | 114 | 79 | |
| GenBank mRNA | BC018431.1 | - | 1 - 2 | 114 | 79 | |
| BY141405.1 | - | 1 - 2 | 131 | 79 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_146248.2 transcripts.
Set Membership: |
|

Cchcr1
Genomic Map