Assay Details
| Gene Symbol: Cchcr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146248.2 | NP_666360.2 | 8 - 9 | 1262 | 102 | |
| GenBank mRNA | AK159780.1 | - | 8 - 9 | 1189 | 102 | |
| BC031416.1 | - | 9 - 10 | 1351 | 4012768 | 102 | |
| CD562711.1 | - | 2 - 3 | 233 | 102 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cchcr1
Genomic Map