Assay Details
| Gene Symbol: Capn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301250.1 | NP_001288179.1 | 1 - 2 | 67 | 59 | |
| GenBank mRNA | BC014767.1 | - | 1 - 2 | 59 | 4458488 | 59 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC014767.1, NM_001301250.1 transcripts.
Set Membership: |
|

Capn5
Genomic Map