Assay Details
| Gene Symbol: Kctd14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC024588.1 | - | 1 - 2 | 93 | 5066294 | 85 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC024588.1 transcripts.
Set Membership: |
|

Kctd14
Genomic Map