| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700033119.mm.1 |
NM_001253885.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700033119.mm.1 |
NM_001253885.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700033119.mm.1 |
NM_001253886.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700033119.mm.1 |
NM_001253886.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700033119.mm.1 |
NM_001253887.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700033119.mm.1 |
NM_001253887.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700033119.mm.1 |
NM_009685.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700033119.mm.1 |
NM_009685.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700033119.mm.1 |
TC0700003981.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700033117.mm.1 |
NM_001253885.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700033117.mm.1 |
NM_001253885.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700033117.mm.1 |
NM_001253886.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700033117.mm.1 |
NM_001253886.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700033117.mm.1 |
NM_001253887.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700033117.mm.1 |
NM_001253887.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700033117.mm.1 |
NM_009685.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700033117.mm.1 |
NM_009685.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700033117.mm.1 |
TC0700003981.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700033118.mm.1 |
NM_001253885.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700033118.mm.1 |
NM_001253885.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700033118.mm.1 |
NM_001253886.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700033118.mm.1 |
NM_001253886.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700033118.mm.1 |
NM_001253887.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700033118.mm.1 |
NM_001253887.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700033118.mm.1 |
NM_009685.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700033118.mm.1 |
NM_009685.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700033118.mm.1 |
TC0700003981.mm.1 |
MTA TC |