Assay Details
| Gene Symbol: Ucp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011671.5 | NP_035801.3 | 3 - 4 | 493 | 82 | |
| GenBank mRNA | AF111998.1 | - | 1 - 2 | 196 | 82 | |
| AF111999.1 | - | 1 - 2 | 190 | 82 | ||
| AK035298.1 | - | 488 | 82 | |||
| AK054060.1 | - | 494 | 82 | |||
| AK134116.1 | - | 3 - 4 | 494 | 82 | ||
| AK142687.1 | - | 3 - 4 | 494 | 82 | ||
| AK165245.1 | - | 3 - 4 | 494 | 82 | ||
| BC012697.1 | - | 3 - 4 | 492 | 4205625 | 82 | |
| BC012967.1 | - | 2 - 3 | 394 | 2811421 | 82 | |
| U69135.1 | - | 3 - 4 | 421 | 82 | ||
| U94593.1 | - | 3 - 4 | 414 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ucp2
Genomic Map