Assay Details
| Gene Symbol: Inppl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122739.1 | NP_001116211.1 | 26 - 27 | 3189 | 211 | |
| NM_010567.2 | NP_034697.2 | 26 - 27 | 3423 | 211 | ||
| GenBank mRNA | AF162781.1 | - | 26 - 27 | 3273 | 211 | |
| AK133359.1 | - | 26 - 27 | 3150 | 211 | ||
| AK149873.1 | - | 26 - 27 | 3206 | 210 | ||
| BC039207.1 | - | 13 - 14 | 1325 | 211 | ||
| BC063080.1 | - | 26 - 27 | 3119 | 6416946 | 211 | |
| U92477.1 | - | 7 - 8 | 775 | 211 | ||
More Information
Set Membership: |
|

Inppl1
Genomic Map