Assay Details
| Gene Symbol: Rrp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133951.1 | NP_598712.1 | 1 - 2 | 318 | 72 | |
| GenBank mRNA | AK154337.1 | - | 1 - 2 | 311 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rrp8
Genomic Map