Assay Details
| Gene Symbol: Tpp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009906.5 | NP_034036.1 | 11 - 12 | 1447 | 113 | |
| GenBank mRNA | AF111172.1 | - | 11 - 12 | 1455 | 113 | |
| AJ011912.1 | - | 11 - 12 | 1441 | 113 | ||
| AK002418.1 | - | 11 - 12 | 1466 | 113 | ||
| AK048279.1 | - | 11 - 12 | 1448 | 113 | ||
| AK083832.1 | - | 10 - 11 | 2712 | 113 | ||
| AK146759.1 | - | 11 - 12 | 1441 | 113 | ||
| AK155021.1 | - | 11 - 12 | 1466 | 113 | ||
| AK165669.1 | - | 11 - 12 | 1473 | 114 | ||
| AK166648.1 | - | 9 - 10 | 1240 | 113 | ||
| AK169924.1 | - | 11 - 12 | 1454 | 113 | ||
| AK170781.1 | - | 11 - 12 | 1443 | 113 | ||
| BC024820.1 | - | 11 - 12 | 1449 | 5360558 | 113 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tpp1
Genomic Map