Assay Details
| Gene Symbol: Tpp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009906.5 | NP_034036.1 | 4 - 5 | 403 | 84 | |
| GenBank mRNA | AF111172.1 | - | 4 - 5 | 411 | 84 | |
| AJ011912.1 | - | 4 - 5 | 397 | 84 | ||
| AK002418.1 | - | 4 - 5 | 422 | 84 | ||
| AK048279.1 | - | 4 - 5 | 404 | 84 | ||
| AK083832.1 | - | 3 - 4 | 1668 | 84 | ||
| AK146759.1 | - | 4 - 5 | 396 | 84 | ||
| AK155021.1 | - | 4 - 5 | 422 | 84 | ||
| AK165669.1 | - | 4 - 5 | 428 | 84 | ||
| AK166648.1 | - | 2 - 3 | 197 | 84 | ||
| AK169924.1 | - | 4 - 5 | 410 | 84 | ||
| AK170781.1 | - | 4 - 5 | 399 | 84 | ||
| AK212650.1 | - | 1 - 2 | 59 | 84 | ||
| BC024820.1 | - | 4 - 5 | 405 | 5360558 | 84 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK083832.1 transcripts.
Set Membership: |
|

Tpp1
Genomic Map