Assay Details
| Gene Symbol: Tpp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009906.5 | NP_034036.1 | 6 - 7 | 701 | 73 | |
| GenBank mRNA | AF111172.1 | - | 6 - 7 | 709 | 73 | |
| AJ011912.1 | - | 6 - 7 | 695 | 73 | ||
| AK002418.1 | - | 6 - 7 | 720 | 73 | ||
| AK048279.1 | - | 6 - 7 | 702 | 73 | ||
| AK083832.1 | - | 5 - 6 | 1966 | 73 | ||
| AK146759.1 | - | 6 - 7 | 694 | 73 | ||
| AK155021.1 | - | 6 - 7 | 720 | 73 | ||
| AK165669.1 | - | 6 - 7 | 726 | 73 | ||
| AK166648.1 | - | 4 - 5 | 495 | 73 | ||
| AK170781.1 | - | 6 - 7 | 697 | 73 | ||
| BC024820.1 | - | 6 - 7 | 703 | 5360558 | 73 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK083832.1 transcripts.
Set Membership: |
|

Tpp1
Genomic Map