Assay Details
| Gene Symbol: Tpp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009906.5 | NP_034036.1 | 8 - 9 | 1089 | 101 | |
| GenBank mRNA | AF111172.1 | - | 8 - 9 | 1097 | 101 | |
| AJ011912.1 | - | 8 - 9 | 1083 | 101 | ||
| AK002418.1 | - | 8 - 9 | 1108 | 101 | ||
| AK048279.1 | - | 8 - 9 | 1090 | 101 | ||
| AK083832.1 | - | 7 - 8 | 2354 | 101 | ||
| AK146759.1 | - | 8 - 9 | 1083 | 101 | ||
| AK155021.1 | - | 8 - 9 | 1108 | 101 | ||
| AK165669.1 | - | 8 - 9 | 1114 | 101 | ||
| AK166648.1 | - | 6 - 7 | 882 | 101 | ||
| AK169924.1 | - | 8 - 9 | 1096 | 101 | ||
| AK170781.1 | - | 8 - 9 | 1085 | 101 | ||
| BC024820.1 | - | 8 - 9 | 1091 | 5360558 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tpp1
Genomic Map