Assay Details
| Gene Symbol: Art5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291354.1 | NP_001278283.1 | 1 - 2 | 395 | 73 | |
| NR_111935.1 | 1 - 2 | 395 | 73 | |||
| GenBank mRNA | BC144826.1 | - | 1 - 2 | 207 | 9053374 | 73 |
| BC144836.1 | - | 1 - 2 | 207 | 73 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC144826.1, NM_001291354.1 transcripts.
Set Membership: |
|

Art5
Genomic Map