Assay Details
| Gene Symbol: Eif3f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025344.2 | NP_079620.2 | 4 - 5 | 681 | 88 | |
| GenBank mRNA | AK002778.1 | - | 4 - 5 | 680 | 88 | |
| AK003372.1 | - | 4 - 5 | 679 | 88 | ||
| AK010333.1 | - | 4 - 5 | 675 | 87 | ||
| AK088537.1 | - | 4 - 5 | 681 | 88 | ||
| AK136797.1 | - | 680 | 88 | |||
| AK152037.1 | - | 4 - 5 | 682 | 88 | ||
| AK186458.1 | - | 2 - 3 | 167 | 88 | ||
| AK192917.1 | - | 3 - 4 | 275 | 88 | ||
| AK197956.1 | - | 4 - 5 | 328 | 88 | ||
| AK200417.1 | - | 2 - 3 | 167 | 88 | ||
| AK209599.1 | - | 2 - 3 | 165 | 88 | ||
| AK214434.1 | - | 2 - 3 | 167 | 88 | ||
| AK217928.1 | - | 3 - 4 | 299 | 88 | ||
| BC070473.1 | - | 4 - 5 | 663 | 30634857 | 88 | |
| BC083190.1 | - | 4 - 5 | 666 | 30358339 | 88 | |
| EF437360.1 | - | 1 - 2 | 89 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Eif3f
Genomic Map