Assay Details
| Gene Symbol: Insc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173767.3 | NP_776128.2 | 11 - 12 | 1681 | 74 | |
| GenBank mRNA | AK014452.1 | - | 10 - 11 | 1443 | 74 | |
| AK050143.1 | - | 8 - 9 | 1176 | 74 | ||
| BC026405.1 | - | 11 - 12 | 1568 | 4211657 | 74 | |
| BC089586.1 | - | 8 - 9 | 1184 | 74 | ||
| BC139196.1 | - | 8 - 9 | 1069 | 8862218 | 74 | |
| BC139197.1 | - | 8 - 9 | 1069 | 8862219 | 74 | |
| DQ205645.1 | - | 10 - 11 | 1394 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Insc
Genomic Map