Assay Details
| Gene Symbol: Insc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173767.3 | NP_776128.2 | 4 - 5 | 747 | 90 | |
| GenBank mRNA | AK014452.1 | - | 3 - 4 | 508 | 90 | |
| AK050143.1 | - | 1 - 2 | 242 | 90 | ||
| BC026405.1 | - | 4 - 5 | 634 | 4211657 | 90 | |
| BC089586.1 | - | 1 - 2 | 251 | 90 | ||
| BC139196.1 | - | 1 - 2 | 135 | 8862218 | 90 | |
| BC139197.1 | - | 1 - 2 | 135 | 8862219 | 90 | |
| DQ205645.1 | - | 3 - 4 | 460 | 90 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK050143.1, BC139196.1, BC139197.1 transcripts.
Set Membership: |
|

Insc
Genomic Map