Assay Details
| Gene Symbol: Insc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173767.3 | NP_776128.2 | 1 - 2 | 244 | 82 | |
| GenBank mRNA | BB852659.1 | - | 1 - 2 | 244 | 82 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_173767.3 transcripts.
Set Membership: |
|

Insc
Genomic Map