Assay Details
| Gene Symbol: Sox6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025559.3 | NP_001020730.1 | 5 - 6 | 673 | 150 | |
| NM_001025560.2 | NP_001020731.1 | 5 - 6 | 793 | 150 | ||
| NM_001277326.1 | NP_001264255.1 | 4 - 5 | 648 | 150 | ||
| NM_001277327.1 | NP_001264256.1 | 4 - 5 | 828 | 150 | ||
| NM_001277328.1 | NP_001264257.1 | 3 - 4 | 536 | 150 | ||
| NM_011445.4 | NP_035575.1 | 5 - 6 | 701 | 150 | ||
| GenBank mRNA | AK029696.1 | - | 3 - 4 | 463 | 150 | |
| AK030713.1 | - | 5 - 6 | 794 | 150 | ||
| AK048143.1 | - | 5 - 6 | 674 | 150 | ||
| AK084290.1 | - | 5 - 6 | 832 | 150 | ||
| AK132409.1 | - | 4 - 5 | 614 | 150 | ||
| BC067407.1 | - | 4 - 5 | 825 | 30094413 | 150 | |
| D61689.1 | - | 3 - 4 | 532 | 150 | ||
| U32614.1 | - | 5 - 6 | 705 | 150 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sox6
Genomic Map