Assay Details
| Gene Symbol: Zfp143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009281.3 | NP_033307.2 | 9 - 10 | 912 | 79 | |
| GenBank mRNA | AF011758.2 | - | 861 | 79 | ||
| AK028056.1 | - | 9 - 10 | 905 | 79 | ||
| AK029078.1 | - | 9 - 10 | 904 | 79 | ||
| AK029926.1 | - | 9 - 10 | 1177 | 79 | ||
| BC037658.1 | - | 8 - 9 | 822 | 5356405 | 79 | |
| U29513.1 | - | 3 - 4 | 234 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zfp143
Genomic Map