Assay Details
| Gene Symbol: Gp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK009843.1 | - | 1 - 2 | 109 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK009843.1 transcripts.
Set Membership: |
|

Gp2
Genomic Map