Assay Details
| Gene Symbol: Gp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025989.3 | NP_080265.2 | 5 - 6 | 987 | 84 | |
| GenBank mRNA | AK009323.1 | - | 5 - 6 | 988 | 84 | |
| AK009661.1 | - | 5 - 6 | 988 | 84 | ||
| AK009843.1 | - | 5 - 6 | 941 | 84 | ||
| BC119573.2 | - | 5 - 6 | 982 | 40057995 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gp2
Genomic Map